siRNA Details
| Gene Symbol: Slc24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020090.1 | NP_064475.1 | 1 | 740 | ||
| GenBank mRNA | AF176688.1 | - | 1 | 740 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc24a1| Gene Symbol: Slc24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020090.1 | NP_064475.1 | 1 | 740 | ||
| GenBank mRNA | AF176688.1 | - | 1 | 740 | ||