siRNA Details
| Gene Symbol: Pcp4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126093.2 | NP_001119565.1 | 3 | 624 | ||
| XM_008769760.1 | XP_008767982.1 | 3 | 1465 | |||
| GenBank mRNA | BC086330.1 | - | 3 | 624 | ||
| BC162039.1 | - | 3 | 466 | 7099690 | ||

Pcp4l1
Genomic Map