siRNA Details
| Gene Symbol: Ube2v1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110345.2 | NP_001103815.2 | 4 | 430 | ||
| GenBank mRNA | CO405669.1 | - | 4 | 405 | ||
| Gene Symbol: LOC100912618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751325.3 | XP_003751373.1 | 1 | 393 | ||
| Gene Symbol: LOC679539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751324.3 | XP_003751372.1 | 1 | 393 | ||

Ube2v1
Genomic Map