siRNA Details
| Gene Symbol: LOC678893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750190.3 | XP_003750238.1 | 3 | 629 | ||
| XM_008764871.1 | XP_008763093.1 | 1 | 2556 | |||
| XM_008764872.1 | XP_008763094.1 | 3 | 2826 | |||
| XM_008764873.1 | XP_008763095.1 | 4 | 3351 | |||
| XM_008764874.1 | XP_008763096.1 | 4 | 1016 | |||
| XM_008764875.1 | XP_008763097.1 | 3 | 892 | |||
| XM_008764876.1 | XP_008763098.1 | 3 | 1138 | |||
| XM_008764877.1 | XP_008763099.1 | 4 | 2889 | |||

LOC678893
Genomic Map