siRNA Details
| Gene Symbol: LOC678893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750190.3 | XP_003750238.1 | 3 | 682 | ||
| XM_008764871.1 | XP_008763093.1 | 1 | 2609 | |||
| XM_008764872.1 | XP_008763094.1 | 3 | 2879 | |||
| XM_008764873.1 | XP_008763095.1 | 4 | 3404 | |||
| XM_008764874.1 | XP_008763096.1 | 4 | 1069 | |||
| XM_008764875.1 | XP_008763097.1 | 3 | 945 | |||
| XM_008764876.1 | XP_008763098.1 | 3 | 1191 | |||
| XM_008764877.1 | XP_008763099.1 | 4 | 2942 | |||

LOC678893
Genomic Map