siRNA Details
Gene Symbol: Gpr158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170326.1 | NP_001163797.1 | 11 | 3808 | ||
XM_008771857.1 | XP_008770079.1 | 9 | 2132 | |||
XM_008771858.1 | XP_008770080.1 | 8 | 1967 | |||
XM_008771859.1 | XP_008770081.1 | 9 | 2152 |
