siRNA Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229520.2 | XP_006229582.1 | 20 | 1886 | ||
| XM_006229522.2 | XP_006229584.1 | 20 | 1883 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp185| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229520.2 | XP_006229582.1 | 20 | 1886 | ||
| XM_006229522.2 | XP_006229584.1 | 20 | 1883 | |||