siRNA Details
| Gene Symbol: Zfp185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229520.2 | XP_006229582.1 | 3 | 286 | ||
| XM_006229522.2 | XP_006229584.1 | 3 | 286 | |||
| XM_008773592.1 | XP_008771814.1 | 3 | 288 | |||
| XM_008773593.1 | XP_008771815.1 | 3 | 289 | |||
| XM_008773594.1 | XP_008771816.1 | 3 | 288 | |||

Zfp185
Genomic Map