siRNA Details
| Gene Symbol: Lrrcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100645.2 | NP_001094115.2 | 2 | 215 | ||
| GenBank mRNA | BG671381.1 | - | 2 | 218 | ||
| CB547116.1 | - | 2 | 161 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrcc1| Gene Symbol: Lrrcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100645.2 | NP_001094115.2 | 2 | 215 | ||
| GenBank mRNA | BG671381.1 | - | 2 | 218 | ||
| CB547116.1 | - | 2 | 161 | |||