siRNA Details
| Gene Symbol: Efcab12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749810.3 | XP_003749858.1 | 8, 9 | 2565 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efcab12| Gene Symbol: Efcab12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749810.3 | XP_003749858.1 | 8, 9 | 2565 | ||