siRNA Details
| Gene Symbol: Nxf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773496.1 | XP_008771718.1 | 9, 10 | 936 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxf5| Gene Symbol: Nxf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773496.1 | XP_008771718.1 | 9, 10 | 936 | ||