siRNA Details
Gene Symbol: LOC685187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764882.1 | XP_008763104.1 | 1250 |
Gene Symbol: LOC103690152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764883.1 | XP_008763105.1 | 2 | 1840 |
