siRNA Details
| Gene Symbol: LOC103690152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764883.1 | XP_008763105.1 | 2 | 2384 | ||
| Gene Symbol: LOC685187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764882.1 | XP_008763104.1 | 1794 | |||

LOC103690152
Genomic Map