siRNA Details
| Gene Symbol: Tsfm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276476.1 | NP_001263405.1 | 6 | 640 | ||
| GenBank mRNA | AJ005161.1 | - | 6 | 641 | ||
| FQ220263.1 | - | 6 | 640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsfm| Gene Symbol: Tsfm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276476.1 | NP_001263405.1 | 6 | 640 | ||
| GenBank mRNA | AJ005161.1 | - | 6 | 641 | ||
| FQ220263.1 | - | 6 | 640 | |||