siRNA Details
| Gene Symbol: Arhgap19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749116.3 | XP_003749164.1 | 4 | 515 | ||
| XM_008760475.1 | XP_008758697.1 | 3 | 249 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap19| Gene Symbol: Arhgap19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749116.3 | XP_003749164.1 | 4 | 515 | ||
| XM_008760475.1 | XP_008758697.1 | 3 | 249 | |||