siRNA Details
| Gene Symbol: Ssx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773085.1 | XP_008771307.1 | 5 | 1610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ssx1| Gene Symbol: Ssx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773085.1 | XP_008771307.1 | 5 | 1610 | ||