siRNA Details
| Gene Symbol: Zcchc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257298.2 | XP_006257360.2 | 1 | 1116 | ||
| GenBank mRNA | FQ213725.1 | - | 1 | 337 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc18| Gene Symbol: Zcchc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257298.2 | XP_006257360.2 | 1 | 1116 | ||
| GenBank mRNA | FQ213725.1 | - | 1 | 337 | ||