siRNA Details
| Gene Symbol: Tmem169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109574.1 | NP_001103044.1 | 3 | 508 | ||
| XM_006245284.2 | XP_006245346.1 | 2 | 419 | |||
| XM_006245285.2 | XP_006245347.1 | 2 | 415 | |||
| XM_006245286.2 | XP_006245348.1 | 3 | 441 | |||
| XM_006245287.2 | XP_006245349.1 | 2 | 529 | |||

Tmem169
Genomic Map