siRNA Details
| Gene Symbol: Esx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773498.1 | XP_008771720.1 | 3 | 528 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esx1| Gene Symbol: Esx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773498.1 | XP_008771720.1 | 3 | 528 | ||