siRNA Details
| Gene Symbol: Kmt2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762670.1 | XP_008760892.1 | 4 | 796 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kmt2c| Gene Symbol: Kmt2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762670.1 | XP_008760892.1 | 4 | 796 | ||