siRNA Details
| Gene Symbol: Fam168b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271134.1 | NP_001258063.1 | 3, 4 | 327 | ||
| XM_008766976.1 | XP_008765198.1 | 2, 3 | 921 | |||
| XM_008766977.1 | XP_008765199.1 | 2, 3 | 917 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam168b| Gene Symbol: Fam168b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271134.1 | NP_001258063.1 | 3, 4 | 327 | ||
| XM_008766976.1 | XP_008765198.1 | 2, 3 | 921 | |||
| XM_008766977.1 | XP_008765199.1 | 2, 3 | 917 | |||