siRNA Details
| Gene Symbol: Dnal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109477.2 | NP_001102947.2 | 4 | 247 | ||
| XM_006240352.2 | XP_006240414.1 | 5 | 456 | |||
| GenBank mRNA | BC161941.1 | - | 286 | |||
| EV765393.1 | - | 5 | 286 | |||
| FQ211665.1 | - | 251 | ||||

Dnal1
Genomic Map