siRNA Details
| Gene Symbol: Smim20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134639.2 | NP_001128111.2 | 3 | 725 | ||
| GenBank mRNA | FQ215060.1 | - | 3 | 719 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim20| Gene Symbol: Smim20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134639.2 | NP_001128111.2 | 3 | 725 | ||
| GenBank mRNA | FQ215060.1 | - | 3 | 719 | ||