siRNA Details
| Gene Symbol: LOC689587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253734.2 | XP_006253796.1 | 4 | 1529 | ||
| XM_006253735.2 | XP_006253797.1 | 4 | 1508 | |||
| XM_008771555.1 | XP_008769777.1 | 3 | 1030 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC689587| Gene Symbol: LOC689587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253734.2 | XP_006253796.1 | 4 | 1529 | ||
| XM_006253735.2 | XP_006253797.1 | 4 | 1508 | |||
| XM_008771555.1 | XP_008769777.1 | 3 | 1030 | |||