siRNA Details
| Gene Symbol: Casq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159594.1 | NP_001153066.1 | 8, 9 | 1057 | ||
| GenBank mRNA | FM056035.1 | - | 5, 6 | 362 | ||
| FQ216689.1 | - | 8, 9 | 634 | |||
| FQ224875.1 | - | 9, 10 | 884 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Casq1| Gene Symbol: Casq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159594.1 | NP_001153066.1 | 8, 9 | 1057 | ||
| GenBank mRNA | FM056035.1 | - | 5, 6 | 362 | ||
| FQ216689.1 | - | 8, 9 | 634 | |||
| FQ224875.1 | - | 9, 10 | 884 | |||