siRNA Details
Gene Symbol: LOC688542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760597.1 | XP_008758819.1 | 783 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC688542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760597.1 | XP_008758819.1 | 783 |