siRNA Details
Gene Symbol: Mgarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109556.1 | NP_001103026.1 | 3 | 268 | ||
XM_006232330.2 | XP_006232392.1 | 3 | 224 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mgarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109556.1 | NP_001103026.1 | 3 | 268 | ||
XM_006232330.2 | XP_006232392.1 | 3 | 224 |