siRNA Details
| Gene Symbol: Mgarp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109556.1 | NP_001103026.1 | 4 | 766 | ||
| XM_006232330.2 | XP_006232392.1 | 4 | 722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgarp| Gene Symbol: Mgarp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109556.1 | NP_001103026.1 | 4 | 766 | ||
| XM_006232330.2 | XP_006232392.1 | 4 | 722 | |||