siRNA Details
Gene Symbol: Tspan15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115032.1 | NP_001108504.1 | 3, 4 | 453 | ||
GenBank mRNA | BC158654.1 | - | 3, 4 | 453 | 8368493 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tspan15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115032.1 | NP_001108504.1 | 3, 4 | 453 | ||
GenBank mRNA | BC158654.1 | - | 3, 4 | 453 | 8368493 |