siRNA Details
Gene Symbol: Vrtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750199.3 | XP_003750247.1 | 1 | 1197 | ||
XM_008764891.1 | XP_008763113.1 | 1 | 1197 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vrtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750199.3 | XP_003750247.1 | 1 | 1197 | ||
XM_008764891.1 | XP_008763113.1 | 1 | 1197 |