siRNA Details
| Gene Symbol: Elovl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044275.1 | NP_001037740.1 | 3 | 339 | ||
| GenBank mRNA | BC085795.1 | - | 3 | 339 | 7109632 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elovl1| Gene Symbol: Elovl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044275.1 | NP_001037740.1 | 3 | 339 | ||
| GenBank mRNA | BC085795.1 | - | 3 | 339 | 7109632 | |