siRNA Details
| Gene Symbol: Vom2r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099511.1 | NP_001092981.1 | 3, 4 | 1310 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r42| Gene Symbol: Vom2r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099511.1 | NP_001092981.1 | 3, 4 | 1310 | ||
Set Membership: |
|