siRNA Details
| Gene Symbol: Fam150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109372.1 | NP_001102842.1 | 2 | 718 | ||
| GenBank mRNA | BC167018.1 | - | 2 | 759 | 7307720 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam150b| Gene Symbol: Fam150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109372.1 | NP_001102842.1 | 2 | 718 | ||
| GenBank mRNA | BC167018.1 | - | 2 | 759 | 7307720 | |