siRNA Details
| Gene Symbol: Zfp654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768541.1 | XP_008766763.1 | 5 | 706 | ||
| XM_237855.8 | XP_237855.6 | 5 | 706 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp654| Gene Symbol: Zfp654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768541.1 | XP_008766763.1 | 5 | 706 | ||
| XM_237855.8 | XP_237855.6 | 5 | 706 | |||