siRNA Details
| Gene Symbol: Lpgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109376.2 | NP_001102846.1 | 2 | 388 | ||
| XM_008769875.1 | XP_008768097.1 | 3 | 446 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpgat1| Gene Symbol: Lpgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109376.2 | NP_001102846.1 | 2 | 388 | ||
| XM_008769875.1 | XP_008768097.1 | 3 | 446 | |||