siRNA Details
Gene Symbol: Ndufs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019223.1 | NP_062096.1 | 2 | 283 |
Gene Symbol: LOC100912599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748682.3 | XP_003748730.1 | 3, 4 | 324 |
Gene Symbol: LOC679739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001053017.4 | XP_001053017.1 | 3, 4 | 324 | ||
GenBank mRNA | FQ224183.1 | - | 3, 4 | 331 | ||
FQ224635.1 | - | 4, 5 | 452 |
