siRNA Details
| Gene Symbol: LOC100912599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748682.3 | XP_003748730.1 | 1, 2 | 156 | ||
| Gene Symbol: LOC679739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001053017.4 | XP_001053017.1 | 1, 2 | 156 | ||
| GenBank mRNA | FQ224183.1 | - | 1, 2 | 163 | ||

LOC100912599
Genomic Map