siRNA Details
| Gene Symbol: Tma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126048.2 | NP_001119520.1 | 4 | 311 | ||
| GenBank mRNA | BC157813.1 | - | 4 | 311 | ||
| FQ221914.1 | - | 4 | 287 | |||
| FQ222972.1 | - | 4 | 276 | |||
| Gene Symbol: LOC100910678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749743.3 | XP_003749791.1 | 2 | 287 | ||
| XM_006236476.2 | XP_006236538.1 | 1 | 439 | |||

Tma7
Genomic Map