siRNA Details
| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305043.1 | NP_001291972.1 | 2 | 519 | ||
| NM_001305049.1 | NP_001291978.1 | 2 | 298 | |||
| NM_001305051.1 | NP_001291980.1 | 2 | 298 | |||
| NM_001305066.1 | NP_001291995.1 | 2 | 519 | |||
| NM_004872.4 | NP_004863.2 | 2 | 519 | |||
| GenBank mRNA | AF047439.1 | - | 2 | 283 | ||
| AF290615.1 | - | 2 | 519 | |||
| AK075187.1 | - | 2 | 459 | |||
| AK075505.1 | - | 2 | 282 | |||
| AK123445.1 | - | 1 | 722 | |||
| AK225624.1 | - | 2 | 284 | |||
| AX775907.1 | - | 2 | 519 | |||
| AX775909.1 | - | 2 | 286 | |||
| AY359029.1 | - | 2 | 338 | |||
| BC003106.1 | - | 2 | 268 | 3501639 | ||
| BC016374.1 | - | 2 | 286 | 4618034 | ||
| BC062738.1 | - | 1 | 17 | |||
| BC095464.1 | - | 2 | 311 | 30554552 | ||
| BP371330.1 | - | 2 | 297 | |||
| BU165003.1 | - | 2 | 157 | |||
| BX463615.2 | - | 2 | 156 | |||
| DB459209.1 | - | 2 | 297 | |||

TMEM59
Genomic Map