siRNA Details
| Gene Symbol: Clec5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109377.1 | NP_001102847.1 | 5 | 459 | ||
| XM_006236369.2 | XP_006236431.1 | 4 | 424 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec5a| Gene Symbol: Clec5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109377.1 | NP_001102847.1 | 5 | 459 | ||
| XM_006236369.2 | XP_006236431.1 | 4 | 424 | |||