siRNA Details
| Gene Symbol: Fer1l5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767045.1 | XP_008765267.1 | 53, 54 | 6442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fer1l5| Gene Symbol: Fer1l5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767045.1 | XP_008765267.1 | 53, 54 | 6442 | ||