siRNA Details
| Gene Symbol: Glipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001054584.5 | XP_001054584.2 | 4 | 351 | ||
| XM_006238140.2 | XP_006238202.1 | 5 | 1179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glipr2| Gene Symbol: Glipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001054584.5 | XP_001054584.2 | 4 | 351 | ||
| XM_006238140.2 | XP_006238202.1 | 5 | 1179 | |||