siRNA Details
Gene Symbol: Dnajc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053690.2 | NP_446142.2 | 2 | 624 | ||
XM_006240726.1 | XP_006240788.1 | 2 | 697 | |||
XM_006240727.1 | XP_006240789.1 | 1 | 563 | |||
XM_008765011.1 | XP_008763233.1 | 2 | 686 | |||
GenBank mRNA | AF351783.1 | - | 2 | 678 | ||
BC083549.1 | - | 2 | 624 | 7127738 |
Gene Symbol: LOC498205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769500.1 | XP_008767722.1 | 1 | 580 |
