siRNA Details
| Gene Symbol: AP2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014203.2 | NP_055018.2 | 18 | 2502 | ||
| NM_130787.2 | NP_570603.2 | 17 | 2436 | |||
| XM_011526556.2 | XP_011524858.1 | 18 | 3112 | |||
| XM_011526557.2 | XP_011524859.1 | 17 | 3041 | |||
| GenBank mRNA | AK090661.1 | - | 2 | 1523 | ||
| AK094317.1 | - | 8 | 938 | |||
| AL136925.1 | - | 17 | 2302 | |||
| BC014214.2 | - | 17 | 2334 | 4111165 | ||

AP2A1
Genomic Map