siRNA Details
| Gene Symbol: Slc9b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109385.2 | NP_001102855.1 | 4 | 633 | ||
| GenBank mRNA | BC166815.1 | - | 4 | 633 | 7460062 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9b2| Gene Symbol: Slc9b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109385.2 | NP_001102855.1 | 4 | 633 | ||
| GenBank mRNA | BC166815.1 | - | 4 | 633 | 7460062 | |