siRNA Details
| Gene Symbol: Klhdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034133.1 | NP_001029305.1 | 8, 9 | 943 | ||
| GenBank mRNA | BC105756.1 | - | 8, 9 | 943 | 5623547 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhdc2| Gene Symbol: Klhdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034133.1 | NP_001029305.1 | 8, 9 | 943 | ||
| GenBank mRNA | BC105756.1 | - | 8, 9 | 943 | 5623547 | |