siRNA Details
| Gene Symbol: Tcta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243774.2 | XP_006243836.1 | 1 | 110 | ||
| XM_008766514.1 | XP_008764736.1 | 1 | 110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcta| Gene Symbol: Tcta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243774.2 | XP_006243836.1 | 1 | 110 | ||
| XM_008766514.1 | XP_008764736.1 | 1 | 110 | |||