siRNA Details
| Gene Symbol: Gpr183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109386.1 | NP_001102856.1 | 2 | 1007 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr183| Gene Symbol: Gpr183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109386.1 | NP_001102856.1 | 2 | 1007 | ||
Set Membership: |
|