siRNA Details
| Gene Symbol: Asb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170749.1 | NP_001164220.1 | 8 | 1515 | ||
| XM_008771008.1 | XP_008769230.1 | 9 | 1619 | |||
| XM_008771009.1 | XP_008769231.1 | 3 | 929 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb14| Gene Symbol: Asb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170749.1 | NP_001164220.1 | 8 | 1515 | ||
| XM_008771008.1 | XP_008769230.1 | 9 | 1619 | |||
| XM_008771009.1 | XP_008769231.1 | 3 | 929 | |||