siRNA Details
| Gene Symbol: LOC100360426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773639.1 | XP_008771861.1 | 1 | 492 | ||
| Gene Symbol: Tomm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001055588.5 | XP_001055588.3 | 2 | 432 | ||

LOC100360426
Genomic Map